Learn More
Biocare Medical, LLC p40 (P) Antibody, Recognizes the Shortest Variant of Human p53, 6 ml
SDP
Catalog No. NC0283339
Encompass_Preferred
p40 may be a valuable marker in cases where p63 has been used. p40 may aid in the differential diagnosis of lung ADC vs. lung SqCC. In a study, p40 exhibited markedly superior specificity vs. p63 for SqCC and equivalent sensitivity.
Catalog No. NC0283339 Supplier Biocare Medical, LLC Supplier No. API3030AA
p40 may be a valuable marker in cases where p63 has been used. p40 may aid in the differential diagnosis of lung ADC vs. lung SqCC. In a study, p40 exhibited markedly superior specificity vs. p63 for SqCC and equivalent sensitivity.