Learn More
FARIBO MANUFACTURING COMPANY LDPE Rectangular Mobile Tub with Lid, 28 gal; 1/Pk
Catalog No. NC0675138
Encompass_Preferred
Tubs are made of low-density polyethylene (LDPE). Rugged caster frame supports heavy loads. Three-inch diameter casters make rolling easy. Container and casters can be removed from zinc-plated frame for cleaning. Tubs include polyethylene cover. NSF certified; USDA approved. Capacity (L)106 - Capacity (Gal) 28 - Color White Material LDPE Cap Material Polyethylene (PE) Overall Length (cm) 67.9 Overall Width (cm) 52.1 Overall Height (cm) 42.5
Catalog No. NC0675138 Supplier FARIBO MANUFACTURING COMPANY Supplier No. P433C/A1
Tubs are made of low-density polyethylene (LDPE). Rugged caster frame supports heavy loads. Three-inch diameter casters make rolling easy. Container and casters can be removed from zinc-plated frame for cleaning. Tubs include polyethylene cover. NSF certified; USDA approved. Capacity (L)106 - Capacity (Gal) 28 - Color White Material LDPE Cap Material Polyethylene (PE) Overall Length (cm) 67.9 Overall Width (cm) 52.1 Overall Height (cm) 42.5