Learn More
Arbor Assays P450 Demethylation Activity Kit (2 plate)
SDP
Supplier:  Arbor Assays K011F1
Encompass_Preferred
The DetectX P450 Fluorescent Activity Kit allows activity measurement of any demethylating P450 system WITHOUT additions to the P450:Substrate reaction by measuring the formaldehyde generated by demethylation. Lab-to-lab comparisons are possible with inclusion of a formaldehyde reference standard.
Catalog No. 50-227-8393