Learn More
Electron Microscopy Sciences P47 Scintillator for Novascan, SEMCO/Zeiss, 16.4 x 1 mm
SDP
Supplier:  Electron Microscopy Sciences 82022
Encompass_Preferred
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on Novascan, SEMCO/Zeiss.
Size: 16.4 x 1 mm
Catalog No. 50-276-19