Learn More
Electron Microscopy Sciences P47 Scintillator for Hitachi w/metal ring, 19.8 x 1 mm
SDP
Supplier:  Electron Microscopy Sciences 82029
Encompass_Preferred
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on Hitachi w/metal ring.
Size: 19.8 x 1 mm
Catalog No. 50-276-06