Learn More
Electron Microscopy Sciences P47 Scintillator for JEOL JSM T20, T200 T300, 340, 8.5 x 2 mm
SDP
Supplier:  Electron Microscopy Sciences 82013
Encompass_Preferred
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on JEOL JSM T20, T200 T300, 340.
Size: 8.5 x 2 mm
Catalog No. 50-276-15