Learn More
Electron Microscopy Sciences P47 Scintillator for ISI Mini-SEM, 7.5 x 1 mm
SDP
Catalog No. 5027616
Encompass_Preferred
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on ISI Mini-SEM.
Size: 7.5 x 1 mm
Catalog No. 50-276-16 Supplier Electron Microscopy Sciences Supplier No. 82012
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on ISI Mini-SEM.
Size: 7.5 x 1 mm