Learn More
Electron Microscopy Sciences P47 Scintillator for for JEOL U3, JSM2, 19.0 x 1 mm
SDP
Catalog No. 5027609
Encompass_Preferred
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on JEOL U3, JSM2.
Size: 19.0 x 1 mm
Catalog No. 50-276-09 Supplier Electron Microscopy Sciences Supplier No. 82026
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on JEOL U3, JSM2.
Size: 19.0 x 1 mm