Learn More
Electron Microscopy Sciences P47 Scintillator for Philips SEM, 20.0 x 2 mm
SDP
Supplier:  Electron Microscopy Sciences 82032
Encompass_Preferred
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on Philips SEM.
Size: 20.0 x 2 mm
Catalog No. 50-276-04