Learn More
Electron Microscopy Sciences P47 Scintillator for Zeiss SEM, 12.0 x 1 mm
SDP
Catalog No. 5027611
Encompass_Preferred
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on Zeiss SEM.
Size: 12.0 x 1 mm
Catalog No. 50-276-11 Supplier Electron Microscopy Sciences Supplier No. 82018
They are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required. Use on Zeiss SEM.
Size: 12.0 x 1 mm