Learn More
Enzo Life Sciences p47Phox polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIOSA5500200
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human p47Phox. UniProt ID: P14598. Source: Purified from rabbit serum. Species reactivity: Human, Mouse. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7372