Learn More
Aviva Systems Biology Corp P4HA2 Antibody - C-terminal region (ARP62190_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62190_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against P4HA2. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-605