Learn More
Epigentek Inc P4HA3 Polyclonal Antibody, FITC Conjulated (100 ul)
SDP
Supplier:  Epigentek Inc A57319100
Encompass_Preferred
Biotin Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Human Prolyl 4-hydroxylase subunit alpha-3 protein (20-544AA). Tested Applications: ELISA. Reactivity: Human. Alternative Names: Procollagen-proline, 2-oxoglutarate-4-dioxygenase subunit alpha-3, UNQ711/PRO1374, P4HA3.
Catalog No. 50-122-8732