Learn More
Aviva Systems Biology Corp P4HB Antibody - N-terminal region (ARP48150_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48150_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against P4HB. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human P4HB. Alternate names: DSI, ERBA2L, GIT, PDI, PDIA1, PHDB, PO4DB, PO4HB, PROHB, P4HbetaApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59732