Learn More
Medchemexpress LLC P4pal10 TFA | 98.25% | 1409.72 | 5 MG
SDP
Catalog No. 5000397905
Encompass_Preferred
P4pal10 TFA is the TFA salt form of P4pal10. It acts as an antagonist for protease-activated receptor 4 (PAR4). It inhibits platelet aggregation, tissue factor (TF)-induced thrombin generation, and demonstrates anticoagulant and antithrombotic activities. Additionally, it reduces edema and granulocyte infiltration induced by Carrageenan and ameliorates injury in rats with myocardial ischemia/reperfusion (I/R) models.
  • Antagonist for protease-activated receptor 4 (PAR4)
  • Inhibits platelet aggregation
  • Inhibits tissue factor (TF)-induced thrombin generation
  • Demonstrates anticoagulant and antithrombotic activities
  • Reduces edema and granulocyte infiltration
  • Ameliorates injury in myocardial ischemia/reperfusion (I/R) models
Catalog No. 50-003-97905 Supplier Medchemexpress LLC Supplier No. HYP5875A5MG
P4pal10 TFA is the TFA salt form of P4pal10. It acts as an antagonist for protease-activated receptor 4 (PAR4). It inhibits platelet aggregation, tissue factor (TF)-induced thrombin generation, and demonstrates anticoagulant and antithrombotic activities. Additionally, it reduces edema and granulocyte infiltration induced by Carrageenan and ameliorates injury in rats with myocardial ischemia/reperfusion (I/R) models.
  • Antagonist for protease-activated receptor 4 (PAR4)
  • Inhibits platelet aggregation
  • Inhibits tissue factor (TF)-induced thrombin generation
  • Demonstrates anticoagulant and antithrombotic activities
  • Reduces edema and granulocyte infiltration
  • Ameliorates injury in myocardial ischemia/reperfusion (I/R) models