Learn More
Medchemexpress LLC P5(PEG24)-VC-PAB-exatecan | 2928571-43-9 | 99.74% | 2105.28 | 5 MG
SDP
Catalog No. 5000392378
Encompass_Preferred
P5(PEG24)-VC-PAB-Exatecan (LP5) is a drug-linker conjugate that combines a potent DNA Topoisomerase I inhibitor, Exatecan, with a P5(PEG24)-VC-PAB linker to form an antibody-drug conjugate (ADC). It is intended for tumor research.
  • Drug-linker conjugate for ADC
  • Composed of a potent DNA Topoisomerase I inhibitor exatecan
  • Can be used for the research of tumor
  • For research use only
Catalog No. 50-003-92378 Supplier Medchemexpress LLC Supplier No. HY1630995MG
P5(PEG24)-VC-PAB-Exatecan (LP5) is a drug-linker conjugate that combines a potent DNA Topoisomerase I inhibitor, Exatecan, with a P5(PEG24)-VC-PAB linker to form an antibody-drug conjugate (ADC). It is intended for tumor research.
  • Drug-linker conjugate for ADC
  • Composed of a potent DNA Topoisomerase I inhibitor exatecan
  • Can be used for the research of tumor
  • For research use only