Learn More
Werther International P50/24 SILENT COMPRESSOR 115V
SDP
Catalog No. 501037799
Encompass_Preferred
Silent air compressor, P50/24AL 115V/60Hz, Output 2.1 CFM / 60 l.min, Max Pressure 120 PSI, Noise Level 40 dB/A, 6 Gal air tank, 2 year warranty, Dimensions 17 in.L x 17 in.W x 20 in.H
Catalog No. 50-103-7799 Supplier Werther International Supplier No. M06090US
Silent air compressor, P50/24AL 115V/60Hz, Output 2.1 CFM / 60 l.min, Max Pressure 120 PSI, Noise Level 40 dB/A, 6 Gal air tank, 2 year warranty, Dimensions 17 in.L x 17 in.W x 20 in.H