Learn More
Biocare Medical, LLC P504S Antibody - UltraLine, 25 ml For Roche Benchmark Ultra
SDP
Supplier:  Biocare Medical, LLC AVA200G25
Encompass_Preferred
P504S, also known as α-methylacyl coenzyme A racemase (AMACR), is a peroxisomal and mitochondrial enzyme that plays a role in bile acid synthesis and β-oxidation of branched chain fatty acids. P504S was initially identified from a cDNA library as a gene that is overexpressed in human prostate cancer; with little or no expression in normal prostate. In immunohistochemistry, P504S (P) antibody has been shown to be a specific marker of prostatic adenocarcinoma. Additionally, prostate glands involved in PIN have been found to express P504S, whereas P504S was nearly undetectable in benign glands.
Catalog No. 50-833-18