Learn More
Enzo Life Sciences P504S + p63 + HMWCK antibody cocktaIL-(10ml)
SDP
Supplier:  Enzo Life Sciences ENZABS7170010
Encompass_Preferred
Isotype: IgG. Immunogen: A peptide corresponding to residues in human P504S (AMACR), recombinant p63, keratin solubilized from human stratum corneum. Species reactivity: Human. Applications: IHC. Purity: Affinity-purified. Formulation: Liquid. In DPBS buffer, pH 7.4, containing 1% BSA and 0.05% preservative.
Catalog No. 50-201-3257