Learn More
TARGETMOL CHEMICALS INC P505-15 ACETATE 5MG
SDP
Catalog No. 502580796
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. P505-15 Acetate is a selective inhibitor of spleen tyrosine kinase that acts by suppressing leukocyte immune function and inflammation and leading to a reduction in arthritis score and attenuated histological damage. purity: 99%
Catalog No. 50-258-0796 Supplier TARGETMOL CHEMICALS INC Supplier No. T245845MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. P505-15 Acetate is a selective inhibitor of spleen tyrosine kinase that acts by suppressing leukocyte immune function and inflammation and leading to a reduction in arthritis score and attenuated histological damage. purity: 99%