Learn More
Abcam P5091, USP-7 inhibitor, 5MG
Catalog No. 502705579
Encompass_Preferred
MW 348.2 Da, Purity >98%. Potent, selective and cell-permeable USP-7 inhibitor (EC₅₀ = 4.2 μM). Induces apoptosis in cancer cells in vitro. Antiangiogenic and antitumor agent. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5579 Supplier Abcam Supplier No. AB1446025MG
MW 348.2 Da, Purity >98%. Potent, selective and cell-permeable USP-7 inhibitor (EC₅₀ = 4.2 μM). Induces apoptosis in cancer cells in vitro. Antiangiogenic and antitumor agent. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement