Learn More
AMPLYUS LLC miniPCR bio P51™ Molecular Fluorescence Viewer
SDP
Catalog No. NC1812433
Encompass_Preferred
Make biology glow! With the P51 Fluorescence Viewer, you can detect and measure molecules like DNA, RNA, and proteins in a simple, visual way, without the sophisticated equipment traditional fluorescence detection requires.
Catalog No. NC1812433 Supplier AMPLYUS LLC Supplier No. QP190001
Make biology glow! With the P51 Fluorescence Viewer, you can detect and measure molecules like DNA, RNA, and proteins in a simple, visual way, without the sophisticated equipment traditional fluorescence detection requires.