Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P53 (17-26), FITC labeled | 98.3% | 1752.98 | 10 MG
SDP

Catalog No. 5000423801
Encompass_Preferred

P53 (17-26), FITC labeled is a biologically active peptide intended for research use. It presents as a light yellow to orange solid with a purity of 98.31%.

  • For research use only
  • Molecular weight: 1752.98
  • Appearance: Solid
  • Color: Light yellow to orange
  • Sequence: {FITC-Ahx}-Glu-Thr-Phe-Ser-Asp-Leu-Trp-Lys-Leu-Leu-NH2
  • Soluble in DMSO (≥ 100 mg/mL)

Catalog No. 50-004-23801 Supplier Medchemexpress LLC Supplier No. HYP1755F10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P53 (17-26), FITC labeled is a biologically active peptide intended for research use. It presents as a light yellow to orange solid with a purity of 98.31%.

  • For research use only
  • Molecular weight: 1752.98
  • Appearance: Solid
  • Color: Light yellow to orange
  • Sequence: {FITC-Ahx}-Glu-Thr-Phe-Ser-Asp-Leu-Trp-Lys-Leu-Leu-NH2
  • Soluble in DMSO (≥ 100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.