Learn More
Biogenex Laboratories p53 : 1801 : 6mL
SDP
Supplier:  Biogenex Laboratories AM2405M
Encompass_Preferred
p53 is a tumor suppressor gene product identified in a wide variety oftumors. p53 protein accumulates within the malignant cells where it can bereadily detected by immunohistochemistry. p53 protein is present in lowconcentrations in normal cells, but elevated levels of mutant p53 havebeen found in many common cancers. Accumulation of mutant p53detected by immunohistochemical staining has been reported in breast,lung, colon, stomach, bladder, and testis carcinomas, soft-tissue sarcomas,and melanomas.
Catalog No. 50-197-8826