Learn More
Medchemexpress LLC p53 Activator 5 | 2636840-37-2 | 98.43% | 566.58 | 5 MG
SDP
Catalog No. 5000362243
Encompass_Preferred
p53 Activator 5 (compound 134A) is a potent p53 activator that can bind to mutant p53 and restore its ability to bind DNA. This compound exhibits anti-tumor activity and is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent p53 activator with an SC150 value of <0.05 mM
  • Binds to mutant p53 and restores DNA binding ability
  • Exhibits anti-tumor activity
  • Click chemistry reagent with an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-62243 Supplier Medchemexpress LLC Supplier No. HY1484045MG
p53 Activator 5 (compound 134A) is a potent p53 activator that can bind to mutant p53 and restore its ability to bind DNA. This compound exhibits anti-tumor activity and is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent p53 activator with an SC150 value of <0.05 mM
  • Binds to mutant p53 and restores DNA binding ability
  • Exhibits anti-tumor activity
  • Click chemistry reagent with an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups