Learn More
CPC Scientific H-Gly-Ser-Arg-Ala-His-Ser-Ser-His-Leu-Lys-Ser-Lys-Lys-Gly-Gln-Ser-Thr-Ser-Arg-His-Lys-Lys-Trp-Lys-Met-Arg-Arg-Asn-Gln-Phe-Trp-Val-Lys-Val-Gln-Arg-Gly-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CASP025A
Encompass
ONE-LETTER SEQUENCE: GSRAHSSHLKSKKGQSTSRHKKWKMRRNQFWVKVQRGMOLECULAR FORMULA: C192H315N71O49S1MOLECULAR WEIGHT:4434.2STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: G. Selivanova, et al., Mol. Cell Biol. 19, 3395 (1999); G. Selivanova, et al., Nat. Med. 3, 632 (1997); G. Selivanova, et al., Nucleic Acids Res. 24, 3560 (1996)
Catalog No. 50-210-8965