Learn More
Biomatik Corporation P53 Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50245
Encompass_Preferred
P53 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Tumor Protein P53. The P53 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: 7~393aa. Applications: ELISA, WB, IHC, IF. Target Synonyms: Tumor Protein P53. Accession Number: P04637. Gene ID: 7157. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7727
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.