Learn More
Santa Cruz Biotechnology p53 Antibody (Pab 240) | 200 µg/ml
Supplier:  Santa Cruz Biotechnology SC99
Encompass
p53 Antibody (Pab 240) is a mouse monoclonal IgG1 κ, cited in 439 publications, provided at 200 µg/ml. Raised against amino acids 14-389 of p53 of human origin. p53 Antibody (Pab 240) is recommended for detection of only mutant p53 under non-denaturing conditions but is equally reactive with mutant and wildtype p53 under denaturing conditions of mouse, rat, human and origin by WB, IP and IF; also reactive with additional species, including bovine and avian. Anti-p53 Antibody (Pab 240) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM.
Catalog No. NC1825391