Learn More
Biogenex Laboratories p53 : DO7 : 6mL
SDP
Supplier:  Biogenex Laboratories AM2395M
Encompass_Preferred
p53 is a tumor suppressor gene product identified in a wide variety oftumors. p53 protein is present in low concentration in normal cells, butelevated levels of mutant p53 have been found in many common cancers.Accumulation of mutant p53 detected by immunohistochemical staininghas been reported in breast, lung, colon, stomach, bladder, and testiscarcinomas, soft-tissue sarcomas, and melanomas.
Catalog No. 50-197-8825