Learn More
Enzo Life Sciences p53 (human), (recombinant) (GST-tag) (50µg)
SDP
Supplier:  Enzo Life Sciences BMLFW93700050
Encompass_Preferred
Alternative name: TRP53, Tumor protein p53. Formulation: Liquid. In 20 mM Tris-HCl, 0.15 M NaCl, pH 8.0. Source: Produced in E. coli. Full length human p53 is fused to a GST-tag. Long Term Storage: -80°C.
Catalog No. 50-201-0841