Learn More
Enzo Life Sciences p53 (human), (recombinant) (His-tag) (20µg)
SDP
Supplier:  Enzo Life Sciences BMLFW88200020
Encompass_Preferred
Alternative name: TRP53, Tumor protein p53. Formulation: Liquid. In 20 mM Tris-Cl (pH 8.0), 20% Glycerol, 100 mM KCl, 0.2 mM EDTA, and 1 mM DTT. Source: Produced in insect cells using a baculovirus system. Full length human p53 is fused to a His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-0840