Learn More
Enzo Life Sciences p53 (pSer315) monoclonal antibody (FPS315) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC239E
Encompass_Preferred
Clone: FPS315. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of human p53 phosphorylated at Ser315. UniProt ID: P04637. Species reactivity: Human. Applications: WB. Recommended Dilutions: Western Blot (5µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7248