Learn More
Enzo Life Sciences p53 monoclonal antibody (PAb122) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC002D
Encompass_Preferred
Clone: pAb122. Host: Mouse. Isotype: IgG2b. Immunogen: SV40 transformed 3T3 cells. UniProt ID: P02340. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat. Creossreactivity: Weakly cross-reacts with dog p53. Applications: Flow Cytometry, ICC, IHC, IP, WB. Recommended Dilutions: Western Blot (1:250 colorimetric, 1:250 ECL) Immunohistochemistry (1:20 - 1:100) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7231