Learn More
Medchemexpress LLC P53 and MDM2 proteins-interaction-inhibitor (chiral) | 939981-37-0 | 98.1% | C40H49Cl2N5O4 | 5 MG
SDP
Catalog No. 5000434300
Encompass_Preferred
The p53 and MDM2 proteins-interaction-inhibitor (chiral), also known as Compound 32, functions as an inhibitor of the interaction between p53 and MDM2 proteins. This product is intended for research use only and is not suitable for patient administration.
  • Inhibits p53 and MDM2 protein interaction.
  • Suitable for laboratory research.
  • Stable under recommended storage conditions.
Catalog No. 50-004-34300 Supplier Medchemexpress LLC Supplier No. HY700275MG
The p53 and MDM2 proteins-interaction-inhibitor (chiral), also known as Compound 32, functions as an inhibitor of the interaction between p53 and MDM2 proteins. This product is intended for research use only and is not suitable for patient administration.
  • Inhibits p53 and MDM2 protein interaction.
  • Suitable for laboratory research.
  • Stable under recommended storage conditions.