Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P53 and MDM2 proteins-interaction-inhibitor (chiral) | 939981-37-0 | 98.1% | C40H49Cl2N5O4 | 50 MG
SDP

Catalog No. 5000433637
Encompass_Preferred

The p53 and MDM2 proteins-interaction-inhibitor (chiral) is a high-purity chemical compound with a purity of 98.1%. This compound, identified by its chemical formula C40H49Cl2N5O4, functions as an inhibitor of the interaction between p53 and MDM2 proteins. It is supplied as a white to off-white solid, with specific storage requirements for both powder and solution forms to maintain its stability and efficacy.

  • Purity: 98.1%
  • Chemical formula: C40H49Cl2N5O4
  • Molecular weight: 734.75
  • Appearance: White to off-white solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Solubility in vitro: DMSO: 50 mg/mL (68.05 mM)

Catalog No. 50-004-33637 Supplier Medchemexpress LLC Supplier No. HY7002750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The p53 and MDM2 proteins-interaction-inhibitor (chiral) is a high-purity chemical compound with a purity of 98.1%. This compound, identified by its chemical formula C40H49Cl2N5O4, functions as an inhibitor of the interaction between p53 and MDM2 proteins. It is supplied as a white to off-white solid, with specific storage requirements for both powder and solution forms to maintain its stability and efficacy.

  • Purity: 98.1%
  • Chemical formula: C40H49Cl2N5O4
  • Molecular weight: 734.75
  • Appearance: White to off-white solid
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Solubility in vitro: DMSO: 50 mg/mL (68.05 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.