Learn More
Medchemexpress LLC P53 and MDM2 proteins-interaction-inhibitor dihydrochloride | 2070009-27-5 | 99.56% | C40H51Cl4N5O4 | 5 MG
SDP
Catalog No. 5000434721
Encompass_Preferred
P53 and MDM2 proteins-interaction-inhibitor dihydrochloride is an inhibitor of the interaction between p53 and MDM2 proteins. This compound is a chemical entity that modulates biological processes by inhibiting specific protein-protein interactions, making it suitable for various research applications.
  • Inhibits interaction between p53 and MDM2 proteins
  • Used in cancer research
  • Applicable in studies of cancer metabolism and metastasis
  • Relevant for apoptosis research
  • Modulates MDM-2/p53 pathways
Catalog No. 50-004-34721 Supplier Medchemexpress LLC Supplier No. HY70027A5MG
P53 and MDM2 proteins-interaction-inhibitor dihydrochloride is an inhibitor of the interaction between p53 and MDM2 proteins. This compound is a chemical entity that modulates biological processes by inhibiting specific protein-protein interactions, making it suitable for various research applications.
  • Inhibits interaction between p53 and MDM2 proteins
  • Used in cancer research
  • Applicable in studies of cancer metabolism and metastasis
  • Relevant for apoptosis research
  • Modulates MDM-2/p53 pathways