Learn More
Sigma Aldrich Fine Chemicals Biosciences p53 Protein 10 181g Ful
Catalog No. 502908978
Encompass
p53 is expressed in a wide variety of tissues and plays a major role in preventing tumour development. It is involved in cell cycle regulation functioning as a trans-activator to negatively regulate cell division by controlling a set of genes required for this process. It acts as a tumour suppressor in many tumour types by stimulating either apoptosis or growth arrest depending on the physiological circumstances and the cell type. Defects in p53 expression have been implicated in several diseases including choroid plexus papilloma lung cancer head/neck squamous cell carcinomas oesophageal squamous cell carcinoma Li-Fraumeni syndrome and hereditary adrenocortical carcinoma.
Catalog No. 50-290-8978 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 23034M
p53 is expressed in a wide variety of tissues and plays a major role in preventing tumour development. It is involved in cell cycle regulation functioning as a trans-activator to negatively regulate cell division by controlling a set of genes required for this process. It acts as a tumour suppressor in many tumour types by stimulating either apoptosis or growth arrest depending on the physiological circumstances and the cell type. Defects in p53 expression have been implicated in several diseases including choroid plexus papilloma lung cancer head/neck squamous cell carcinomas oesophageal squamous cell carcinoma Li-Fraumeni syndrome and hereditary adrenocortical carcinoma.