Learn More
Medchemexpress LLC p53 Rat His 10ug
SDP
Catalog No. 5000196239
Encompass_Preferred
Recombinant full-length rat p53 protein (M1-D391), expressed in Escherichia coli and supplied lyophilized with an N-terminal 6xHis tag. The protein is formulated for stability and provided at >90% purity for use in biochemical assays, antibody validation, protein-protein interaction studies, and assay development. Store frozen and follow the datasheet for reconstitution and handling to preserve activity.
  • Full-length sequence M1-D391, matching accession P10361.
  • N-terminal 6xHis tag for purification and detection.
  • Expressed in Escherichia coli for high-yield production.
  • Lyophilized formulation with stabilizing excipients for storage stability.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Suitable for biochemical assays, antibody validation, and interaction studies.
  • Recommended to aliquot and store at -20°C or -80°C to minimize freeze-thaw.
Catalog No. 50-001-96239 Supplier Medchemexpress LLC Supplier No. HYP7170310UG
Recombinant full-length rat p53 protein (M1-D391), expressed in Escherichia coli and supplied lyophilized with an N-terminal 6xHis tag. The protein is formulated for stability and provided at >90% purity for use in biochemical assays, antibody validation, protein-protein interaction studies, and assay development. Store frozen and follow the datasheet for reconstitution and handling to preserve activity.
  • Full-length sequence M1-D391, matching accession P10361.
  • N-terminal 6xHis tag for purification and detection.
  • Expressed in Escherichia coli for high-yield production.
  • Lyophilized formulation with stabilizing excipients for storage stability.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Suitable for biochemical assays, antibody validation, and interaction studies.
  • Recommended to aliquot and store at -20°C or -80°C to minimize freeze-thaw.