Learn More
CPC Scientific Biotin-LC-Gly-Ser-Arg-Ala-His-Ser-Ser-His-Leu-Lys-Ser-Lys-Lys-Gly-Gln-Ser-Thr-Ser-Arg-His-Lys-Lys-Leu-Met-Phe-Lys-Thr-Glu-Gly-Pro-Asp-Ser-Asp-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANC020A
Encompass
This peptide is amino acids 361 to 393 fragment of p53 tumor suppressor protein. This sequence is biotinylated through the LC spacer on the N-terminus. p53 is a transcription factor that regulates cell cycle and functions as a tumor supressor. ONE-LETTER SEQUENCE: Biotin-LC-GSRAHSSHLKSKKGQSTSRHKKLMFKTEGPDSD-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:3992.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Cancer
Catalog No. 50-210-8897