Learn More
Biocare Medical, LLC p53 Tumor Suppressor Protein Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM042C
Encompass_Preferred
p53 has been observed to act as both as a tumor-suppressor and transcription factor. p53 activation by DNA damage or other stress signals is reported to trigger DNA repair, cell-cycle arrest, or apoptosis. The nuclear p53 gene is located on chromosome 17p, a frequent site of allele loss in many tumors (60%) including breast, colon and lung. p53 Tumor Suppressor Protein antibody mouse monoclonal has also been shown to have prognostic utility for distal colorectal cancer and nasopharyngeal carcinoma by the assessment of mutation and overexpression status.
Catalog No. 50-824-48