Learn More
Cayman Chemical p53 (cynomolgus monkey) Monoclonal Antibody (Clone 06), 1 mL
SDP
Catalog No. 502914953
Encompass_Preferred
Immunogen: Recombinant cynomolgus monkey p53 • Host: Mouse• Species Reactivity: (+) Cynomolgus monkey • Cross Reactivity: (+) p53 • Applications: ELISA, WB
Catalog No. 50-291-4953 Supplier Cayman Chemical Supplier No. 370691 ML
Immunogen: Recombinant cynomolgus monkey p53 • Host: Mouse• Species Reactivity: (+) Cynomolgus monkey • Cross Reactivity: (+) p53 • Applications: ELISA, WB