Learn More
Cayman Chemical p53cynomolgus monkey Monc 50uL
SDP
Catalog No. 502914955
Encompass_Preferred
A phenol with diverse biological activities is a gut microbiota-derived active metabolite of ferulic acid inhibits a-glucosidase a-amylase and tyrosinase (Kis 47.7 189 and 40.3 Ug/ml respectively) induces cell cycle arrest at the S and G2/M phases and apoptosis in HT-29 cells at 1 mM
Catalog No. 50-291-4955 Supplier Cayman Chemical Supplier No. 3706950 UL
A phenol with diverse biological activities is a gut microbiota-derived active metabolite of ferulic acid inhibits a-glucosidase a-amylase and tyrosinase (Kis 47.7 189 and 40.3 Ug/ml respectively) induces cell cycle arrest at the S and G2/M phases and apoptosis in HT-29 cells at 1 mM