Learn More
TARGETMOL CHEMICALS INC P53R3 5MG
SDP
Catalog No. 502579963
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. P53R3 is a potent p53 reactivator and restores sequence-specific DNA binding of p53 hot spot mutants including p53 R175H p53 R248W and p53 R273H . P53R3 induces p53-dependent antiproliferative effects with much higher specificity than PRIMA-1. P53R3 enhances the recruitment of wild-type p53 and p53 M237I to several target gene promoters. P53R3 strongly enhances the mRNA total protein and cell surface expression of the death receptor death receptor 5 (DR5) . P53R3 is used for cancer research. purity: 97%
Catalog No. 50-257-9963 Supplier TARGETMOL CHEMICALS INC Supplier No. T410685MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. P53R3 is a potent p53 reactivator and restores sequence-specific DNA binding of p53 hot spot mutants including p53 R175H p53 R248W and p53 R273H . P53R3 induces p53-dependent antiproliferative effects with much higher specificity than PRIMA-1. P53R3 enhances the recruitment of wild-type p53 and p53 M237I to several target gene promoters. P53R3 strongly enhances the mRNA total protein and cell surface expression of the death receptor death receptor 5 (DR5) . P53R3 is used for cancer research. purity: 97%