Learn More
enQuire Bioreagents p57Kip2 Anti-Human, Mouse Monoclonal [Clone 57P06], Size: 100ug
Supplier:  enQuire Bioreagents 1028MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: 57P06. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: LS174T, Raji, HT29, SK-BR3 cells. Colon or Prostate carcinomas. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human p57Kip2 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 57kDa.Chromosome: 11p15.5. Gene Symbol: CDKN1C. Gene ID: 1028. Ensemble: ENSG00000273707. Uniprot: XP_005252789.
Catalog No. 50-188-4569