Learn More
Enzo Life Sciences p62 (human) (recombinant) (GST-tag) (50 µg)
SDP
Catalog No. NC1333920
Encompass_Preferred
Alternative name: Sequestosome-1, EBI3-associated protein of 60 kDa. MW: ~74.4kDa. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 1M PBS containing 100mM GSH and 15% glycerol. Source: Produced in E. coli. Human p62 (1-440 aa) is fused to a N-terminal GST-tag. Use/Stability: Stable for at least 12 months after receipt when stored at -20°C. UniProt: Q13501. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C. Long Term Storage: -20°C
Catalog No. NC1333920 Supplier Enzo Life Sciences Supplier No. ENZPRT1200050
Alternative name: Sequestosome-1, EBI3-associated protein of 60 kDa. MW: ~74.4kDa. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 1M PBS containing 100mM GSH and 15% glycerol. Source: Produced in E. coli. Human p62 (1-440 aa) is fused to a N-terminal GST-tag. Use/Stability: Stable for at least 12 months after receipt when stored at -20°C. UniProt: Q13501. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C. Long Term Storage: -20°C