Learn More
Medchemexpress LLC P62-mediated mitopha 100mg | 1809031-84-2 | 392.15 | C14H9IN4O2 | 100 MG
SDP
Catalog No. 5000229351
Encompass_Preferred
P62-mediated mitophagy inducer (PMI) is a small-molecule activator of p62-mediated mitophagy used as a pharmacological tool to study mitophagy pathways. It activates mitochondrial autophagy without recruiting Parkin or collapsing mitochondrial membrane potential, and it stabilizes Nrf2 and upregulates p62 at active concentrations.
  • Activates mitochondrial autophagy independent of Parkin recruitment.
  • Remains active in cells lacking a fully functional PINK1/Parkin pathway.
  • Stabilizes Nrf2 and upregulates p62 expression at effective concentrations.
  • Well characterized by NMR, HRMS, and elemental analysis for research applications.
  • Solid compound, soluble in DMSO for stock preparation.
Catalog No. 50-002-29351 Supplier Medchemexpress LLC Supplier No. HY115576100MG
P62-mediated mitophagy inducer (PMI) is a small-molecule activator of p62-mediated mitophagy used as a pharmacological tool to study mitophagy pathways. It activates mitochondrial autophagy without recruiting Parkin or collapsing mitochondrial membrane potential, and it stabilizes Nrf2 and upregulates p62 at active concentrations.
  • Activates mitochondrial autophagy independent of Parkin recruitment.
  • Remains active in cells lacking a fully functional PINK1/Parkin pathway.
  • Stabilizes Nrf2 and upregulates p62 expression at effective concentrations.
  • Well characterized by NMR, HRMS, and elemental analysis for research applications.
  • Solid compound, soluble in DMSO for stock preparation.