Learn More
Medchemexpress LLC P62-mediated mitopha 10mM 1mL | 1809031-84-2 | 392.15 g·mol⁻1 | C14H9IN4O2 | 1 ML
SDP
Catalog No. 5000228031
Encompass_Preferred
P62-mediated mitophagy inducer is a small-molecule research reagent that activates p62-mediated mitophagy without recruiting Parkin or collapsing mitochondrial membrane potential. It is supplied as a 10 mM stock solution (1 mL) for use in cellular and biochemical studies of selective mitochondrial clearance and autophagy pathways.
  • Activates p62-mediated mitophagy without Parkin recruitment.
  • Provided as a ready-to-use 10 mM stock solution in 1 mL.
  • Suitable for cellular and biochemical assays investigating mitophagy and autophagy.
  • Stable storage recommendations: powder at -20°C for up to 3 years; in solvent at -80°C or -20°C as indicated.
  • Comes with chemical identification and characterization data (CAS, formula, molecular weight) in the supplier datasheet.
Catalog No. 50-002-28031 Supplier Medchemexpress LLC Supplier No. HY11557610MM 1ML
P62-mediated mitophagy inducer is a small-molecule research reagent that activates p62-mediated mitophagy without recruiting Parkin or collapsing mitochondrial membrane potential. It is supplied as a 10 mM stock solution (1 mL) for use in cellular and biochemical studies of selective mitochondrial clearance and autophagy pathways.
  • Activates p62-mediated mitophagy without Parkin recruitment.
  • Provided as a ready-to-use 10 mM stock solution in 1 mL.
  • Suitable for cellular and biochemical assays investigating mitophagy and autophagy.
  • Stable storage recommendations: powder at -20°C for up to 3 years; in solvent at -80°C or -20°C as indicated.
  • Comes with chemical identification and characterization data (CAS, formula, molecular weight) in the supplier datasheet.