Learn More
Medchemexpress LLC P62-mediated mitopha 25mg | 1809031-84-2 | 392.15 g/mol | C14H9IN4O2 | 25 MG
SDP
Catalog No. 5000308142
Encompass_Preferred
P62-mediated mitophagy inducer is a small-molecule research compound that selectively activates p62-dependent mitophagy without recruiting Parkin or collapsing mitochondrial membrane potential. It is intended for in vitro and cellular research applications and is not for human or clinical use.
  • Activates p62-dependent mitophagy in cellular models.
  • Does not recruit Parkin or disrupt mitochondrial membrane potential.
  • Solid powder, light yellow to yellow, suitable for laboratory handling.
  • High purity (99.54%) for biochemical and cellular assays.
  • Stable when stored under recommended conditions (powder: -20°C).
Catalog No. 50-003-08142 Supplier Medchemexpress LLC Supplier No. HY11557625MG
P62-mediated mitophagy inducer is a small-molecule research compound that selectively activates p62-dependent mitophagy without recruiting Parkin or collapsing mitochondrial membrane potential. It is intended for in vitro and cellular research applications and is not for human or clinical use.
  • Activates p62-dependent mitophagy in cellular models.
  • Does not recruit Parkin or disrupt mitochondrial membrane potential.
  • Solid powder, light yellow to yellow, suitable for laboratory handling.
  • High purity (99.54%) for biochemical and cellular assays.
  • Stable when stored under recommended conditions (powder: -20°C).