Learn More
Medchemexpress LLC P62-mediated mitopha 50mg | 1809031-84-2 | 392.15 | C14H9IN4O2 | 50 MG
SDP
Catalog No. 5000229387
Encompass_Preferred
P62-mediated mitophagy inducer is a small-molecule activator of p62-dependent mitophagy used in research to stimulate mitochondrial autophagy without recruiting Parkin or collapsing mitochondrial membrane potential.
  • Activates p62-mediated mitophagy without Parkin recruitment.
  • Promotes mitochondrial autophagy while preserving mitochondrial membrane potential.
  • Suitable for autophagy and mitophagy research applications.
  • Powder form with defined cold-storage stability for long-term storage.
  • Available in small research pack sizes for laboratory use.
  • Molecular formula C14H9IN4O2; molecular weight 392.15 g·mol⁻¹.
Catalog No. 50-002-29387 Supplier Medchemexpress LLC Supplier No. HY11557650MG
Add to Cart
Add to Cart
P62-mediated mitophagy inducer is a small-molecule activator of p62-dependent mitophagy used in research to stimulate mitochondrial autophagy without recruiting Parkin or collapsing mitochondrial membrane potential.
  • Activates p62-mediated mitophagy without Parkin recruitment.
  • Promotes mitochondrial autophagy while preserving mitochondrial membrane potential.
  • Suitable for autophagy and mitophagy research applications.
  • Powder form with defined cold-storage stability for long-term storage.
  • Available in small research pack sizes for laboratory use.
  • Molecular formula C14H9IN4O2; molecular weight 392.15 g·mol⁻¹.