Learn More
Medchemexpress LLC HY-113066A 50mg Medchemexpress, Guanosine 5'-diphosphate disodium salt CAS:7415-69-2 Purity:>98%
SDP
Catalog No. 501969222
Encompass_Preferred
Medchemexpress, HY-115576 10mg P62-mediated mitophagy inducer CAS:1809031-84-2 P62-mediated mitophagy inducer is a mitophagy regulator which activates mitophagy without recruiting Parkin or collapsing ΔΨm and retains activity in cells devoid of a fully functional PINK1/Parkin pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9222 Supplier Medchemexpress LLC Supplier No. HY113066A50MG
Medchemexpress, HY-115576 10mg P62-mediated mitophagy inducer CAS:1809031-84-2 P62-mediated mitophagy inducer is a mitophagy regulator which activates mitophagy without recruiting Parkin or collapsing ΔΨm and retains activity in cells devoid of a fully functional PINK1/Parkin pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.